siRNA Details
| Gene Symbol: Slco3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038643.1 | NP_001033732.1 | 1 | 294 | ||
| NM_023908.2 | NP_076397.2 | 1 | 294 | |||
| GenBank mRNA | AF226324.1 | - | 1 | 192 | ||
| AK031280.1 | - | 1 | 294 | |||
| AK037938.1 | - | 1 | 109 | |||
| AK151781.1 | - | 1 | 264 | |||
| AK152488.1 | - | 1 | 128 | |||
| AK166266.1 | - | 1 | 65 | |||
| BC025059.1 | - | 1 | 127 | 2598913 | ||
| BC094464.1 | - | 1 | 86 | |||

Slco3a1
Genomic Map