siRNA Details
| Gene Symbol: Ern1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023913.2 | NP_076402.1 | 10 | 1122 | ||
| GenBank mRNA | AB031332.1 | - | 10 | 1122 | ||
| AK018505.1 | - | 10 | 1163 | |||
| AK077041.1 | - | 1143 | ||||
| AK170988.1 | - | 10 | 1151 | |||

Ern1
Genomic Map