siRNA Details
| Gene Symbol: 0610009O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024179.5 | NP_077141.2 | 6, 7 | 780 | ||
| GenBank mRNA | AK002437.1 | - | 6, 7 | 770 | ||
| AK141675.1 | - | 743 | ||||
| AK172900.1 | - | 6, 7 | 705 | |||
| BC002117.1 | - | 6, 7 | 754 | 3490474 | ||

0610009O20Rik
Genomic Map