siRNA Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289524.1 | NP_001276453.1 | 4 | 553 | ||
| NM_001289525.1 | NP_001276454.1 | 2 | 300 | |||
| NM_024194.6 | NP_077156.2 | 4 | 556 | |||
| GenBank mRNA | AK011726.1 | - | 4 | 537 | ||
| AK017814.1 | - | 4 | 525 | |||
| AK034978.1 | - | 536 | ||||
| AK148748.1 | - | 4 | 515 | |||
| AK152244.1 | - | 2 | 281 | |||
| AK163288.1 | - | 4 | 524 | |||
| AK170052.1 | - | 4 | 533 | |||

Lrrc40
Genomic Map