siRNA Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024203.3 | NP_077165.2 | 6 | 2225 | ||
| GenBank mRNA | AK005957.1 | - | 3 | 331 | ||
| AK016568.1 | - | 6 | 2191 | |||
| AK132211.1 | - | 6 | 2203 | |||
| AK180201.1 | - | 2 | 156 | |||
| AK187583.1 | - | 3 | 298 | |||
| BC003200.1 | - | 5 | 1128 | |||
| BC066088.1 | - | 6 | 2270 | |||
| BC085163.1 | - | 6 | 3491 | 30618952 | ||
| DQ873694.1 | - | 6 | 2206 | |||

Fam120b
Genomic Map