siRNA Details
| Gene Symbol: Stt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024222.2 | NP_077184.2 | 13 | 2313 | ||
| GenBank mRNA | AK012153.1 | - | 13 | 1983 | ||
| AK018758.2 | - | 13 | 2023 | |||
| AK145674.1 | - | 13 | 1954 | |||
| AK148065.1 | - | 13 | 2012 | |||
| AK152899.1 | - | 13 | 2361 | |||
| AK154979.1 | - | 13 | 2002 | |||
| AK170079.1 | - | 13 | 2025 | |||
| AX799084.1 | - | 13 | 2024 | |||
| BC003206.1 | - | 8 | 1025 | |||
| BC013054.1 | - | 3 | 431 | |||
| BC052433.1 | - | 13 | 2313 | 6837097 | ||

Stt3b
Genomic Map