siRNA Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024239.2 | NP_077201.1 | 5 | 497 | ||
| GenBank mRNA | AB010123.1 | - | 5 | 444 | ||
| AK017600.1 | - | 5 | 452 | |||
| AK019907.1 | - | 5 | 447 | |||
| AK046995.1 | - | 5 | 449 | |||
| AK136961.1 | - | 5 | 1048 | |||
| AK139391.1 | - | 5 | 472 | |||
| AK168810.1 | - | 5 | 470 | |||
| AK220541.1 | - | 3 | 6728 | |||
| BC003497.1 | - | 5 | 440 | 3153829 | ||
| BC006939.1 | - | 5 | 495 | 2649718 | ||
| BC025111.1 | - | 5 | 439 | 4194332 | ||

Stambp
Genomic Map