siRNA Details
| Gene Symbol: Retnlb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023881.4 | NP_076370.3 | 4 | 577 | ||
| GenBank mRNA | AF290871.1 | - | 4 | 579 | ||
| AF323083.1 | - | 4 | 464 | |||
| AK020240.1 | - | 4 | 465 | |||
| BC022650.1 | - | 4 | 447 | 4216606 | ||

Retnlb
Genomic Map