siRNA Details
| Gene Symbol: Asb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023906.3 | NP_076395.2 | 10 | 1776 | ||
| GenBank mRNA | AK010315.1 | - | 10 | 1712 | ||
| AK028489.1 | - | 11 | 1845 | |||
| AK143580.1 | - | 10 | 1711 | |||
| BC023086.1 | - | 10 | 1710 | 4947610 | ||

Asb3
Genomic Map