siRNA Details
| Gene Symbol: Scyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023912.2 | NP_076401.1 | 17 | 2413 | ||
| GenBank mRNA | AF276514.1 | - | 17 | 2413 | ||
| AK019240.1 | - | 16 | 2485 | |||
| AK144134.1 | - | 17 | 2399 | |||
| AK145869.1 | - | 17 | 2433 | |||
| BC034519.1 | - | 17 | 2399 | 3672524 | ||
| HM856325.1 | - | 17 | 2362 | |||
| HM856326.1 | - | 16 | 2492 | |||

Scyl1
Genomic Map