siRNA Details
| Gene Symbol: Drap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291080.1 | NP_001278009.1 | 7 | 983 | ||
| NM_024176.2 | NP_077138.1 | 7 | 962 | |||
| GenBank mRNA | AK005445.1 | - | 6 | 697 | ||
| AK010152.1 | - | 7 | 916 | |||
| AK087456.1 | - | 7 | 788 | |||
| AK154251.1 | - | 6 | 742 | |||
| BC002090.1 | - | 7 | 899 | 3488139 | ||

Drap1
Genomic Map