siRNA Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024188.6 | NP_077150.1 | 17 | 1983 | ||
| GenBank mRNA | AB085609.1 | - | 17 | 1725 | ||
| AK010029.1 | - | 4 | 366 | |||
| AK011354.1 | - | 17 | 1663 | |||
| AK078086.1 | - | 17 | 1981 | |||
| AK146158.1 | - | 17 | 1741 | |||
| AK146342.1 | - | 16 | 1626 | |||
| AK151662.1 | - | 17 | 1722 | |||
| AK151695.1 | - | 17 | 1713 | |||
| AK154327.1 | - | 1 | 95 | |||
| AK167681.1 | - | 17 | 1813 | |||
| BC003422.1 | - | 17 | 1687 | 2598824 | ||

Oxct1
Genomic Map