siRNA Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024239.2 | NP_077201.1 | 10 | 2111 | ||
| GenBank mRNA | AB010123.1 | - | 10 | 2058 | ||
| AK017600.1 | - | 10 | 2066 | |||
| AK019907.1 | - | 11 | 1683 | |||
| AK046995.1 | - | 10 | 2062 | |||
| AK139391.1 | - | 10 | 2086 | |||
| AK168810.1 | - | 10 | 2084 | |||
| AK220541.1 | - | 8 | 8342 | |||
| BC003497.1 | - | 10 | 2057 | 3153829 | ||
| BC006939.1 | - | 10 | 2106 | 2649718 | ||
| BC025111.1 | - | 10 | 2050 | 4194332 | ||

Stambp
Genomic Map