siRNA Details
| Gene Symbol: 1500015O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024283.3 | NP_077245.2 | 2, 3 | 418 | ||
| GenBank mRNA | AK018770.1 | - | 2, 3 | 173 | ||
| AK049805.1 | - | 2, 3 | 419 | |||
| BC002254.1 | - | 2, 3 | 351 | 3493553 | ||

1500015O10Rik
Genomic Map