siRNA Details
| Gene Symbol: Rmnd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024288.2 | NP_077250.2 | 2 | 669 | ||
| GenBank mRNA | AK164384.1 | - | 2 | 670 | ||
| BC050876.1 | - | 2 | 520 | 6512227 | ||
| CF748545.1 | - | 2 | 343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rmnd5a| Gene Symbol: Rmnd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024288.2 | NP_077250.2 | 2 | 669 | ||
| GenBank mRNA | AK164384.1 | - | 2 | 670 | ||
| BC050876.1 | - | 2 | 520 | 6512227 | ||
| CF748545.1 | - | 2 | 343 | |||