siRNA Details
| Gene Symbol: Clcnka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146307.1 | NP_001139779.1 | 6 | 647 | ||
| NM_024412.3 | NP_077723.3 | 7 | 651 | |||
| GenBank mRNA | AF124848.1 | - | 6 | 744 | ||
| AY373832.1 | - | 6 | 602 | |||
| BC037077.1 | - | 7 | 655 | 5042736 | ||

Clcnka
Genomic Map