siRNA Details
| Gene Symbol: Mxra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024263.4 | NP_077225.4 | 6 | 1166 | ||
| GenBank mRNA | AB040488.1 | - | 6 | 1086 | ||
| AB052097.1 | - | 6 | 1044 | |||
| AK004732.1 | - | 6 | 1049 | |||
| BC002140.1 | - | 1 | 163 | |||
| BC026438.1 | - | 6 | 1088 | 4239626 | ||
| EU849596.1 | - | 6 | 1004 | |||
| EU849597.1 | - | 6 | 995 | |||

Mxra8
Genomic Map