siRNA Details
| Gene Symbol: Zc3h18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029993.1 | NP_001025164.1 | 2 | 357 | ||
| NM_001029994.1 | NP_001025165.1 | 2 | 357 | |||
| GenBank mRNA | AK017933.1 | - | 2 | 334 | ||
| AK031413.1 | - | 2 | 306 | |||
| AK159881.1 | - | 2 | 293 | |||
| AK166802.1 | - | 2 | 312 | |||
| BC003319.1 | - | 1 | 205 | |||
| BC030495.1 | - | 2 | 337 | 5400494 | ||
| CD348551.1 | - | 2 | 326 | |||

Zc3h18
Genomic Map