siRNA Details
| Gene Symbol: Plekhf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024413.2 | NP_077724.2 | 2 | 438 | ||
| GenBank mRNA | AK143976.1 | - | 427 | |||
| AK157990.1 | - | 2 | 427 | |||
| AK170283.1 | - | 2 | 437 | |||
| AK171398.1 | - | 2 | 427 | |||
| BC002120.1 | - | 1 | 403 | 3491671 | ||

Plekhf1
Genomic Map