siRNA Details
| Gene Symbol: Ublcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024475.5 | NP_077795.2 | 10 | 1018 | ||
| GenBank mRNA | AK043952.1 | - | 10 | 925 | ||
| AK050031.1 | - | 10 | 929 | |||
| AK147744.1 | - | 10 | 966 | |||
| AK153998.1 | - | 10 | 930 | |||
| BC002236.1 | - | 3 | 292 | |||
| BC056652.1 | - | 6 | 519 | |||
| BC083331.1 | - | 8 | 679 | |||
| BC085111.1 | - | 9 | 921 | 30735741 | ||
| BC094051.1 | - | 4 | 324 | |||
| BC106093.1 | - | 9 | 895 | 5098953 | ||

Ublcp1
Genomic Map