siRNA Details
| Gene Symbol: Txnl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025299.3 | NP_079575.1 | 4 | 602 | ||
| GenBank mRNA | AK004039.1 | - | 4 | 559 | ||
| AK007331.1 | - | 4 | 564 | |||
| AK148287.1 | - | 4 | 585 | |||
| AK166497.1 | - | 4 | 578 | |||
| BC031634.1 | - | 4 | 543 | 3662554 | ||

Txnl4a
Genomic Map