siRNA Details
| Gene Symbol: Zc3h18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029993.1 | NP_001025164.1 | 16 | 2709 | ||
| NM_001029994.1 | NP_001025165.1 | 15 | 2637 | |||
| GenBank mRNA | AK031413.1 | - | 16 | 2659 | ||
| AK131189.1 | - | 13 | 1781 | |||
| AK159881.1 | - | 15 | 2579 | |||
| AK165989.1 | - | 14 | 1992 | |||
| AK169952.1 | - | 5 | 3698 | |||
| BC003319.1 | - | 6 | 881 | |||
| BC030495.1 | - | 15 | 2617 | 5400494 | ||
| BC094330.1 | - | 13 | 1790 | |||

Zc3h18
Genomic Map