siRNA Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024282.3 | NP_077244.1 | 5 | 3286 | ||
| GenBank mRNA | AK146492.1 | - | 5 | 3019 | ||
| BC046816.1 | - | 5 | 3128 | 5702476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Desi2| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024282.3 | NP_077244.1 | 5 | 3286 | ||
| GenBank mRNA | AK146492.1 | - | 5 | 3019 | ||
| BC046816.1 | - | 5 | 3128 | 5702476 | ||