siRNA Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014407.1 | - | 2256 | |||
| AK147619.1 | - | 2274 | ||||
| AK149200.1 | - | 2265 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab22a| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014407.1 | - | 2256 | |||
| AK147619.1 | - | 2274 | ||||
| AK149200.1 | - | 2265 | ||||