siRNA Details
| Gene Symbol: Cyp4f16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024442.1 | NP_077762.1 | 13 | 1874 | ||
| GenBank mRNA | AF233646.1 | - | 13 | 1874 | ||
| AK078557.1 | - | 13 | 1935 | |||
| AK083886.1 | - | 12 | 1784 | |||
| AK146232.1 | - | 13 | 1947 | |||
| BC026539.1 | - | 13 | 1906 | 4982322 | ||

Cyp4f16
Genomic Map