siRNA Details
| Gene Symbol: Pdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159730.1 | NP_001153202.1 | 4 | 1050 | ||
| NM_024458.2 | NP_077778.1 | 4 | 1038 | |||
| GenBank mRNA | AK020798.1 | - | 4 | 1037 | ||
| BC012507.1 | - | 4 | 1024 | 4500900 | ||
| BC021920.1 | - | 4 | 1039 | 4506350 | ||
| BC037176.1 | - | 3 | 3153 | |||
| L08075.1 | - | 4 | 1056 | |||

Pdc
Genomic Map