siRNA Details
| Gene Symbol: Ciao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025296.4 | NP_079572.2 | 7 | 2008 | ||
| GenBank mRNA | AK028376.1 | - | 7 | 1934 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ciao1| Gene Symbol: Ciao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025296.4 | NP_079572.2 | 7 | 2008 | ||
| GenBank mRNA | AK028376.1 | - | 7 | 1934 | ||