siRNA Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111272.1 | NP_001104742.1 | 8, 9 | 1120 | ||
| NM_025303.3 | NP_079579.2 | 9, 10 | 1251 | |||
| GenBank mRNA | AF459099.1 | - | 9, 10 | 1201 | ||
| AF459100.1 | - | 8, 9 | 1070 | |||
| AK015087.1 | - | 2, 3 | 200 | |||
| AK031102.1 | - | 9, 10 | 1178 | |||
| AK171076.1 | - | 9, 10 | 1217 | |||
| BC010300.1 | - | 6, 7 | 926 | |||
| BC025118.1 | - | 8, 9 | 1075 | 3670067 | ||

Stau2
Genomic Map