siRNA Details
| Gene Symbol: Tctex1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025329.3 | NP_079605.1 | 1 | 216 | ||
| GenBank mRNA | AK002589.1 | - | 1 | 175 | ||
| AK004191.1 | - | 1 | 144 | |||
| AK007178.1 | - | 1 | 204 | |||
| AK010027.1 | - | 1 | 174 | |||
| AK010464.1 | - | 1 | 172 | |||
| BC053332.1 | - | 1 | 123 | |||
| BC059716.1 | - | 1 | 217 | 6774264 | ||

Tctex1d2
Genomic Map