siRNA Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025382.6 | NP_079658.2 | 10 | 1883 | ||
| GenBank mRNA | AK003528.1 | - | 4 | 399 | ||
| AK014449.1 | - | 2 | 137 | |||
| AY364165.1 | - | 10 | 1668 | |||
| AY846873.1 | - | 10 | 1884 | |||
| AY846874.1 | - | 10 | 1884 | |||
| BC043306.1 | - | 7 | 1315 | |||
| BC054769.1 | - | 10 | 1776 | 6415086 | ||
| BC056944.1 | - | 3 | 237 | |||

Tmem57
Genomic Map