siRNA Details
| Gene Symbol: Prr13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170911.1 | NP_001164382.1 | 4 | 686 | ||
| NM_025385.3 | NP_079661.1 | 5 | 804 | |||
| GenBank mRNA | AK002430.1 | - | 4 | 654 | ||
| AK003850.1 | - | 5 | 801 | |||
| AK008611.1 | - | 3 | 626 | |||
| AK008894.1 | - | 5 | 764 | |||
| AK154583.1 | - | 5 | 764 | |||
| BC016234.1 | - | 4 | 950 | 4503179 | ||

Prr13
Genomic Map