siRNA Details
| Gene Symbol: Pithd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025411.4 | NP_079687.3 | 4 | 520 | ||
| GenBank mRNA | AF400670.1 | - | 4 | 483 | ||
| AK004208.1 | - | 4 | 521 | |||
| AK013877.1 | - | 2 | 183 | |||
| AK050261.1 | - | 4 | 493 | |||
| BC052695.1 | - | 4 | 420 | 5715038 | ||

Pithd1
Genomic Map