siRNA Details
| Gene Symbol: Sf3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025323.2 | NP_079599.1 | 3 | 385 | ||
| GenBank mRNA | AK002375.1 | - | 3 | 433 | ||
| AK003683.1 | - | 3 | 441 | |||
| AK151648.1 | - | 3 | 428 | |||
| BC019535.1 | - | 3 | 385 | 4458983 | ||

Sf3b6
Genomic Map