siRNA Details
| Gene Symbol: Tctex1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025329.3 | NP_079605.1 | 1, 2 | 234 | ||
| GenBank mRNA | AK002589.1 | - | 1, 2 | 193 | ||
| AK004191.1 | - | 1, 2 | 162 | |||
| AK007178.1 | - | 1, 2 | 222 | |||
| AK010027.1 | - | 1, 2 | 192 | |||
| AK010464.1 | - | 1, 2 | 190 | |||
| BC059716.1 | - | 1, 2 | 235 | 6774264 | ||

Tctex1d2
Genomic Map