siRNA Details
| Gene Symbol: Sdhaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025333.4 | NP_079609.2 | 4 | 722 | ||
| GenBank mRNA | AK013454.1 | - | 4 | 627 | ||
| AK075594.1 | - | 4 | 627 | |||
| AK160175.1 | - | 2 | 355 | |||
| AK160957.1 | - | 4 | 629 | |||
| AK167228.1 | - | 4 | 658 | |||
| BC029630.1 | - | 3 | 441 | |||
| BC038924.1 | - | 3 | 690 | |||
| BC048913.1 | - | 4 | 455 | |||
| BC085277.1 | - | 4 | 624 | 30441476 | ||

Sdhaf2
Genomic Map