siRNA Details
| Gene Symbol: Chchd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025336.1 | NP_079612.1 | 3 | 311 | ||
| GenBank mRNA | AK002263.1 | - | 3 | 376 | ||
| AK002889.1 | - | 3 | 383 | |||
| AK003519.1 | - | 3 | 311 | |||
| AK006643.1 | - | 3 | 377 | |||
| AK145337.1 | - | 3 | 360 | |||
| AK193067.1 | - | 2 | 137 | |||
| AK213116.1 | - | 2 | 137 | |||
| AK217481.1 | - | 2 | 137 | |||
| BC016528.1 | - | 2 | 133 | |||
| BC021941.1 | - | 3 | 363 | 5124504 | ||
| BC032987.1 | - | 3 | 326 | |||

Chchd3
Genomic Map