siRNA Details
| Gene Symbol: Akr7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025337.3 | NP_079613.3 | 2, 3 | 529 | ||
| GenBank mRNA | AF525358.2 | - | 2, 3 | 431 | ||
| AJ271800.1 | - | 2, 3 | 446 | |||
| AJ271801.1 | - | 2, 3 | 446 | |||
| AK002664.1 | - | 2, 3 | 509 | |||
| AK003915.1 | - | 2, 3 | 516 | |||
| AK143203.1 | - | 2, 3 | 528 | |||
| BC031857.1 | - | 2, 3 | 506 | 5124841 | ||

Akr7a5
Genomic Map