siRNA Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025364.2 | NP_079640.1 | 3, 4 | 356 | ||
| GenBank mRNA | AK003453.1 | - | 3, 4 | 208 | ||
| AK018773.1 | - | 3, 4 | 219 | |||
| AK088266.1 | - | 3, 4 | 205 | |||
| AK146248.1 | - | 3, 4 | 234 | |||
| AK212934.1 | - | 4, 5 | 240 | |||
| AW824990.1 | - | 3, 4 | 363 | |||
| BC027510.1 | - | 3, 4 | 195 | 1382354 | ||
| BC094393.1 | - | 3, 4 | 192 | |||
| BC157909.1 | - | 3, 4 | 220 | 9088111 | ||
| BC158023.1 | - | 3, 4 | 220 | 9007329 | ||
| BC172063.1 | - | 3, 4 | 217 | 9054349 | ||

Sarnp
Genomic Map