siRNA Details
| Gene Symbol: Josd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205070.1 | NP_001191999.1 | 5 | 741 | ||
| NM_001205071.1 | NP_001192000.1 | 4 | 629 | |||
| NM_001205072.1 | NP_001192001.1 | 4 | 598 | |||
| NM_001205073.1 | NP_001192002.1 | 4 | 595 | |||
| NM_025368.4 | NP_079644.1 | 4 | 626 | |||
| GenBank mRNA | AK002872.1 | - | 4 | 467 | ||
| AK003525.1 | - | 4 | 551 | |||
| BC018244.1 | - | 5 | 824 | 4224952 | ||
| CA326008.1 | - | 4 | 540 | |||

Josd2
Genomic Map