siRNA Details
| Gene Symbol: Dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025384.3 | NP_079660.1 | 3, 4 | 238 | ||
| GenBank mRNA | AK002598.1 | - | 3, 4 | 234 | ||
| AK003396.1 | - | 3, 4 | 248 | |||
| AK007806.1 | - | 3, 4 | 227 | |||
| AK007833.1 | - | 3, 4 | 227 | |||
| BC027509.1 | - | 3, 4 | 238 | 1364626 | ||

Dnajc15
Genomic Map