siRNA Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164472.1 | NP_001157944.1 | 4 | 503 | ||
| NM_025391.2 | NP_079667.2 | 5 | 644 | |||
| GenBank mRNA | AK003739.1 | - | 5 | 644 | ||
| AK006057.1 | - | 5 | 623 | |||
| AK014290.1 | - | 5 | 603 | |||
| BC003972.1 | - | 5 | 631 | 3488726 | ||

Nip7
Genomic Map