siRNA Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025408.2 | NP_079684.2 | 2 | 245 | ||
| GenBank mRNA | AK004287.1 | - | 2 | 246 | ||
| AK011668.1 | - | 2 | 226 | |||
| AK017361.1 | - | 150 | ||||
| AK080977.1 | - | 2 | 223 | |||
| AK150629.1 | - | 2 | 206 | |||
| AK162403.1 | - | 2 | 261 | |||
| BC023924.1 | - | 2 | 249 | 5324078 | ||

Acer3
Genomic Map