siRNA Details
| Gene Symbol: Ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025409.3 | NP_079685.1 | 2 | 239 | ||
| GenBank mRNA | AK004281.1 | - | 2 | 224 | ||
| AK007388.1 | - | 2 | 187 | |||
| AK007862.1 | - | 2 | 171 | |||
| AK009116.1 | - | 2 | 172 | |||
| AK190140.1 | - | 2 | 110 | |||
| BC021512.1 | - | 2 | 229 | 5351700 | ||
| CX207811.1 | - | 2 | 228 | |||

Ier3ip1
Genomic Map