siRNA Details
| Gene Symbol: Myo19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025414.3 | NP_079690.2 | 18, 19 | 2335 | ||
| GenBank mRNA | AK018609.1 | - | 3, 4 | 365 | ||
| AK080117.1 | - | 16, 17 | 2054 | |||
| BC007156.1 | - | 11, 12 | 1251 | 3486558 | ||
| BC046294.1 | - | 17, 18 | 2063 | |||
| BC060115.1 | - | 16, 17 | 2188 | |||

Myo19
Genomic Map