siRNA Details
| Gene Symbol: 0610012G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002600.1 | - | 1 | 612 | ||
| AK002881.1 | - | 1 | 577 | |||
| AK009191.1 | - | 1 | 619 | |||
| AK011122.1 | - | 1 | 606 | |||
| AK146135.1 | - | 1 | 633 | |||
| BC021536.1 | - | 1 | 611 | |||

0610012G03Rik
Genomic Map