siRNA Details
| Gene Symbol: Tmem167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025335.3 | NP_079611.2 | 4 | 2788 | ||
| GenBank mRNA | AK012161.1 | - | 4 | 2722 | ||
| AK031706.1 | - | 4 | 2723 | |||
| AK088895.1 | - | 4 | 2708 | |||
| AK132361.1 | - | 4 | 2708 | |||
| AK161037.1 | - | 4 | 2725 | |||

Tmem167
Genomic Map