siRNA Details
| Gene Symbol: Tmed3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025360.2 | NP_079636.2 | 3 | 895 | ||
| GenBank mRNA | AK002698.1 | - | 3 | 843 | ||
| AK004545.1 | - | 3 | 843 | |||
| AK007544.1 | - | 3 | 837 | |||
| BC023338.1 | - | 3 | 789 | 5253777 | ||

Tmed3
Genomic Map