siRNA Details
| Gene Symbol: Tmem8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159602.1 | NP_001153074.1 | 5 | 741 | ||
| NM_025376.3 | NP_079652.1 | 5 | 893 | |||
| GenBank mRNA | AK003298.1 | - | 5 | 895 | ||
| AK004477.1 | - | 5 | 861 | |||
| BC062145.1 | - | 5 | 819 | 30286808 | ||

Tmem8c
Genomic Map