siRNA Details
| Gene Symbol: Ska2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025377.3 | NP_079653.1 | 3 | 163 | ||
| GenBank mRNA | AK003196.1 | - | 3 | 191 | ||
| AK013317.1 | - | 3 | 164 | |||
| AK013370.1 | - | 3 | 164 | |||
| AK017811.1 | - | 3 | 164 | |||
| AK153219.1 | - | 134 | ||||
| AK162628.1 | - | 160 | ||||
| BC052151.1 | - | 164 | ||||
| BC060088.1 | - | 166 | ||||
| BC096627.1 | - | 175 | ||||
| BC117001.1 | - | 3 | 193 | 40126320 | ||
| BC119167.1 | - | 3 | 193 | 8733916 | ||

Ska2
Genomic Map