siRNA Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025382.6 | NP_079658.2 | 11 | 3425 | ||
| GenBank mRNA | AK014449.1 | - | 3 | 1680 | ||
| AY364165.1 | - | 11 | 3210 | |||
| AY846873.1 | - | 11 | 3427 | |||
| BC043306.1 | - | 8 | 2857 | |||
| BC056944.1 | - | 3 | 3730 | |||

Tmem57
Genomic Map